Ice scoop, stainless steel
Supplier: Sampling Systems
A314-100
SAMPA314-100EA 0 SGD
SAMPA314-100
Ice scoop, stainless steel
Scoops
The ice scoop is designed for scooping ice cubes and pieces of broken ice.
  • 304 stainless steel
  • Smooth handle
  • Robust design
  • Easy to use
The body of the scoop has many 3 mm perforations to allow any melt water to easily drain away.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904