Scoops, Scorpion PharmaScoop®
Supplier: Sampling Systems
A346-1500
SAMPA346-1500EA 0 SGD
SAMPA346-1500
Scoops, Scorpion PharmaScoop®
Scoops
These heavy duty sampling scoops are made of 316 stainless steel with satin finish.
  • Ergonomic handle makes scooping easy when space is tight as well as keeping hands clean
  • Hygienic crevice free construction
  • Radiused corners for easy cleaning
The fully welded construction makes these powder scoops ideal for heavy duty or hygienic applications.