Square scoops, PharmaScoops®
Supplier: Sampling Systems
A366-1
SAMPA366-1EA 0 SGD
SAMPA366-1
Square scoops, PharmaScoops®
Scoops
The square scoops made from 316 stainless steel are ideal for dispensing and sampling powders and granules along flat surfaces.
  • Heavy duty construction
  • Hygienic crevice free construction
  • Satin finish
  • Radiused corners for easy cleaning
Order Now
SPECIFICATIONS
Environmentally Preferable