X-ray and metal detectable volumetric spoons
Supplier: Sampling Systems
M1030B-25
SAMPM1030B-25EA 0 SGD
SAMPM1030B-25
X-ray and metal detectable volumetric spoons
Spoons
The X-ray and metal detectable volumetric spoons are perfect for use in the laboratory and production.
  • Robust PP construction
  • PP gives excellent chemical resistance to many products
  • Supplied individually bagged and produced in a cleanroom
  • Storage: Dry and ambient temperature
  • Full batch traceability
The 2,5 ml spoon size is perfect for measuring small amounts. The measuring spoons are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.More Product Information