Programmable magnetic stirrer, MagStir Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-0302
SCIISI-0302EA 0 SGD
SCIISI-0302
Programmable magnetic stirrer, MagStir Genie®
Stirrers Magnetic Stirrers
Programmable magnetic stirrer with wide range of applications including stirring of cell cultures and viscous solutions.
  • Low profile stirrer with precise and reproducible control
  • Three power levels; low, medium and high - depending on viscosity of the sample
  • Manual mode provides stirring-, reverse direction- and time (1 second to 99 hours) functions
  • Profiles mode allows to program speed, ramp rate, stirring direction, time (including pause), and power level in up to nine distinct steps
  • Automatic mode recalls and executes saved programs
  • Up to four stirring
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Speed range 10 - 1800 min⁻¹
5 - 300 min⁻¹ (ramp rate)
Max. stirring capacity H₂O 2000 ml
Weight 1,0 kg
W×D×H 160×200×55 mm