Hydrometer according to Œchsle
Supplier: Ludwig Schneider GmbH & CO. KG
3002596
SCNL3002596EA 0 SGD
SCNL3002596
Hydrometer according to Œchsle
Hydrometers
High precision instrument for use as test medium in the quality management of the food industry.
  • High quality, high safety and reliability
  • Calibration in accredited lab (optional)
  • Easy to read scales, breakage protected
Especially used in the cider industry.
Certifications: HACCP.
Packaging: Each packed in a square plastic sleeve.