Digital measuring devices, PHYSICS 100
Supplier: Ludwig Schneider GmbH & CO. KG
63874 63731
SCNL63874EA 0 SGD
SCNL63874 SCNL63731
Digital measuring devices, PHYSICS 100
Monitoring Systems CO2 and Temperature Monitoring Systems
PHYSICS 100 provides precise measurements with a wide range of measuring units and sensor technologies. It is suitable for measuring temperature, air humidity, air flow, pressure, flow rate, electrical values.
  • Compact shape and ergonomic design
  • Intelligent probe display with probe specific functions
  • Language selection: English, German or French
  • With graphic display, 128×64 pixel, 7/16 lines with 2 white LEDs
It can have one or two input probes and the main measuring functions involve measuring value, zero-setting, max./min. value documentation, 2-stepMore Product Information