Digital measuring devices, PHYSICS 300
Supplier: Ludwig Schneider GmbH & CO. KG
63885 63884
SCNL63885EA 0 SGD
SCNL63885 SCNL63884
Digital measuring devices, PHYSICS 300
Monitoring Systems CO2 and Temperature Monitoring Systems
PHYSICS 300 provides a wide range of measuring units and sensor technologies with data logger function. It is suitable for measuring temperature, air humidity, air flow, pressure, flow rate, electrical values etc. With its data logger functions, it provides precise measurements in meteorology, water analystics, chemistry, biotechnology, industry, research and development etc. Three measuring points can be covered simultaneously.
  • Compact shape and ergonomic design
  • Intelligent probe display with probe specific functions
  • With three input probes for 65
More Product Information