pH combination electrodes, ScienceLine
Supplier: SI Analytics
285100001 285100034 285100042 285100207 285100223 285101260
SCOI285100001EA 0 SGD
SCOI285100001 SCOI285100034 SCOI285100042 SCOI285100207 SCOI285100223 SCOI285101260
pH combination electrodes, ScienceLine
Electrodes pH Electrodes
Heavy-duty combination electrodes for practically all applications in the laboratory and in the field.
  • Maximum measuring accuracy and consistency, with optimum sensor service life
  • Maximum adaptability for all applications, even at high temperatures
  • Enable measurements in deep vessels with a length of up to 600 mm, in MMR tubes with a diameter of 3 mm, and in round bottom flask with NS 14,5 standard taper
* Without cable (available separately).
Electrolyte: KCl 3 mol/l
Shaft material: Glass
Temperature range: −5 to +100 °C (H 61 / H&More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable