Bio process sterilisable pH and ORP electrodes, SteamLine
Supplier: SI Analytics
285113213 285113246
SCOI285113213EA 0 SGD
SCOI285113213 SCOI285113246
Bio process sterilisable pH and ORP electrodes, SteamLine
Electrodes pH Electrodes
Autoclavable high-tech electrodes with outstanding electrical and physical performance characteristics guaranteeing exceptional accuracy and reliability over long periods of time. Can be used in demanding environments such as SIP treatment (Sterilisation in Place), and CIP cleaning (Clean in Place).
  • Low maintenance electrodes
  • Pressurised reference electrode allowing a significant longer operational time of the electrode
  • S-membrane glass avoiding damages from hot caustic soda
  • Allows up to 100 sterilisations with overheated water vapour at about 135 °C
  • For
More Product Information