Buffer solutions
Supplier: SI ANALYTICS
285138238
SCOI285138238EA 0 SGD
SCOI285138238
Buffer solutions
Buffers pH Calibration Buffers
Buffer solutions intended for extremely precise calibration.
  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam
Certifications: According to DIN 19 266.