Titrators, Karl Fischer, coulometric, TitroLine® 7500 KF Trace
Supplier: SI Analytics
285227800
SCOI285227800EA 0 SGD
SCOI285227800
Titrators, Karl Fischer, coulometric, TitroLine® 7500 KF Trace
Titrators
These very compact, easy to operate instruments are ideal for coulometric water determination according to Karl Fischer in low water content, as well as for bromine number determination. They are suitable for small liquid or gaseous sample amounts, and can be used for quality control in the chemical, petrochemical, pharmaceutical and food industries.
  • High visibility, full colour display with online titration curve
  • Standard methods for different applications (for example blank value)
  • Include three USB and two RS232 ports for expansion and connection of
More Product Information