HPLC bottles, DURAN®
Supplier: DWK Life Sciences
1129820 1129821
SCOT1129820EA 283 SGD
SCOT1129820 SCOT1129821
HPLC bottles, DURAN®
Bottles HPLC Bottles
DURAN®, borosilicate glass 3.3, clear, with blue PP screw cap.
  • Connection of different hose diameters (1,6 and 3,2 mm) and sterile pressure equalisation sets possible
  • Unused ports can be sealed with silicone blanking seals
  • Pressure resistant conforms to DIN ISO 1595
Typical applications: Safe transfer of liquid media within a closed and sterile system (evaporation is reduced). With easy to read scale and large labelling field for easy marking. Complete system comprising: DURAN® pressure plus laboratory bottle with four-port screw cap; four screwMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable