Bottles, narrow neck, with standard ground stoppers, DURAN®
Supplier: DWK Life Sciences
211651702 211652407 211653609 211654408 211655404 211656306 211657302 211658607 211659106 211681705 211682401 211683603 211684402 211685407 211686309 211687305 211688601 211689109
SCOT211651702EA 35.8 SGD
SCOT211651702 SCOT211652407 SCOT211653609 SCOT211654408 SCOT211655404 SCOT211656306 SCOT211657302 SCOT211658607 SCOT211659106 SCOT211681705 SCOT211682401 SCOT211683603 SCOT211684402 SCOT211685407 SCOT211686309 SCOT211687305 SCOT211688601 SCOT211689109
Bottles, narrow neck, with standard ground stoppers, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass type I according to USP, EP and JP
  • Amber glass meets USP <671> and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
ISO 4796-2.
With printed Retrace Code (batch identification).
Order Now
SPECIFICATIONS
Environmentally Preferable