Bottles, wide neck, with standard ground joint stoppers, DURAN®
Supplier: DWK Life Sciences
211852403 211853605 211854404 211855409 211856302 211857307 211858603 211859102 211882406 211883608 211884407 211885403 211886305 211887301 211888606 211889105
SCOT211852403EA 44.6 SGD
SCOT211852403 SCOT211853605 SCOT211854404 SCOT211855409 SCOT211856302 SCOT211857307 SCOT211858603 SCOT211859102 SCOT211882406 SCOT211883608 SCOT211884407 SCOT211885403 SCOT211886305 SCOT211887301 SCOT211888606 SCOT211889105
Bottles, wide neck, with standard ground joint stoppers, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
With printed retrace code (batch identification).
ISO 4796-2
Certifications: ISO 4796-2.