Flasks acc. to Kjeldahl, narrow neck, with flared rim, DURAN®
Supplier: DWK Life Sciences
212311707 212312403 212313605 212314404 212315109 212315409
SCOT212311707EA 19.3 SGD
SCOT212311707 SCOT212312403 SCOT212313605 SCOT212314404 SCOT212315109 SCOT212315409
Flasks acc. to Kjeldahl, narrow neck, with flared rim, DURAN®
Flasks Kjeldahl Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.
Certifications: DIN 12360