Bottles, wide neck, clear, with GLS 80® thread and baffles, DURAN®
Supplier: DWK Life Sciences
212863658 212864457 212865453
SCOT212863658EA 30.3 SGD
SCOT212863658 SCOT212864457 SCOT212865453
Bottles, wide neck, clear, with GLS 80® thread and baffles, DURAN®
Bottles Glass Bottles
Wide neck bottles made of clear DURAN® borosilicate glass 3.3, with GLS 80® thread, have three vertical baffles into the inner surface, the swirling motion is disrupted, giving a much more effective mixing motion.
  • Bottles are fully autoclavable
  • Easy to read graduated volume scale
  • Very good temperature resistance
  • Complete with PP pouring ring and GLS 80® screw cap
The baffles improve 'top to bottom' circulation and produce greater radial mixing. The baffles also offer other advantages such as suppressing the formation of a central vortex and reducing 'More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable