Cell culture bottles, Breed-Demeter, square, DURAN®
Supplier: DWK Life Sciences
214813104
SCOT214813104EA 29.1 SGD
SCOT214813104
Cell culture bottles, Breed-Demeter, square, DURAN®
Bottles Cell Culture Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cell and tissue culture.
Order Now
SPECIFICATIONS
Environmentally Preferable