Glass stoppers, standard ground joint, octagonal, DURAN®
Supplier: DWK Life Sciences
216240307 216240401 216241106 216241209 216241303 216241603 216270404 216240804 216240907 216270901 216271306 216240701
SCOT216240307EA 15.4 SGD
SCOT216240307 SCOT216240401 SCOT216241106 SCOT216241209 SCOT216241303 SCOT216241603 SCOT216270404 SCOT216240804 SCOT216240907 SCOT216270901 SCOT216271306 SCOT216240701
Glass stoppers, standard ground joint, octagonal, DURAN®
Stoppers Glass Stoppers
These stoppers are made from borosilicate glass 3.3 and have an octagonal head for good grip.
  • Solid or semi-hollow
Certifications: DIN 12252
Order Now
SPECIFICATIONS
Environmentally Preferable