Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
218011453 218011753 218012458 218013651 218014459 218015455 218017353 218018658 218019157 218018855 218012955 218015155 218016957 218010851 218016357
SCOT218011453EA 7.8 SGD
SCOT218011453 SCOT218011753 SCOT218012458 SCOT218013651 SCOT218014459 SCOT218015455 SCOT218017353 SCOT218018658 SCOT218019157 SCOT218018855 SCOT218012955 SCOT218015155 SCOT218016957 SCOT218010851 SCOT218016357
Laboratory bottles, narrow neck, with screw cap, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear, with PP screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduation, DIN thread, pouring ring and blue screw cap.
With printed Retrace Code (batch identification).
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominal volumeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable