Laboratory bottles, narrow neck, with thread, DURAN® PROTECT
Supplier: DWK Life Sciences
218053655
SCOT218053655EA 31.6 SGD
SCOT218053655
Laboratory bottles, narrow neck, with thread, DURAN® PROTECT
Bottles Glass Bottles
DURAN® PROTECT coated Borosilicate glass 3.3, clear, with or without screw cap.
  • Very good chemical resistance and good heat resistance (max. thermal load 135 °C), constant loads longer than 30 mins should be avoided
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Compressive strength is not increased by the plastic coating
  • Glass Type I according to USP, EP and JP
Round, with graduation and DIN thread. Without pouring ring or screw cap. The coating provides protection against mechanical damage (scratches, etc) andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable