Hydra square bottle, GL 45, with 4 side necks, DURAN®
Supplier: DWK Life Sciences
218505453
SCOT218505453EA 716 SGD
SCOT218505453
Hydra square bottle, GL 45, with 4 side necks, DURAN®
Bottles Glass Bottles
HPLC 1000 ml purging bottle with five GL 45 inlets.
  • Convenient – purging bottle accepts up to five HPLC GL 45 caps
  • Safer – capped bottles reduce the risk of volatile organic compound (VOC) escape and spillage
  • Compatible - GL 45 threads fit standard HPLC connection caps
  • Manufactured from the material of choice for HPLC solvent bottles: Borosilicate 3.3 glass offering a very low inorganic ion extractables profile
  • 1 L square space saving shape
  • Durable white enamel volume graduations (100 to 800 ml, in 100 ml intervals)
DURAN® bottles are ideal for theMore Product Information