YOUTILITY® bottles, glass, clear, with screw cap, DURAN®
Supplier: DWK Life Sciences
218812854 218814452 218813653 218815457
SCOT218812854EA 39 SGD
SCOT218812854 SCOT218814452 SCOT218813653 SCOT218815457
YOUTILITY® bottles, glass, clear, with screw cap, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate 3.3 glass, clear.
  • Specially shaped grip zones on the bottles for safer and easier handling
  • Ergonomic GL 45 screw cap made from food grade polypropylene
  • Shortened threads on bottle and cap for faster opening
  • Dedicated accessories for easy and simple identification and personalisation
  • Glass Type I according to USP, EP and JP
The DURAN® YOUTILITY bottle represents a new generation of glass laboratory bottles for use in a wide and diverse range of applications. The innovations include improved ergonomic bottle shape that extends toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable