YOUTILITY® bottles, amber, glass, with screw cap, DURAN®
Supplier: DWK Life Sciences
218862859 218863658 218864457 218865453
SCOT218862859EA 144 SGD
SCOT218862859 SCOT218863658 SCOT218864457 SCOT218865453
YOUTILITY® bottles, amber, glass, with screw cap, DURAN®
Bottles Glass Bottles
DURAN®, Borosilicate 3.3 glass, amber.
  • Amber coloured to protect light-sensitive materials
  • Specially shaped grip zones on the bottles for safer and easier handling
  • Ergonomic GL 45 screw cap made from food grade polypropylene
  • Shortened threads on bottle and cap for faster opening
  • Dedicated accessories for easy and simple identification and personalisation
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
The DURAN® YOUTILITY bottle represents a new generation of glass laboratoryMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable