TILT bottles, DURAN®
Supplier: DWK Life Sciences
218914459
SCOT218914459EA 78.8 SGD
SCOT218914459
TILT bottles, DURAN®
Bottles Cell Culture Bottles
The TILT bottle, made of clear DURAN® borosilicate glass 3.3, and is ideal for cell culture media preparation.
  • Low bottle height for easy handling in biosafety cabinets
  • Space saving shape for fridge storage and water baths
  • 45° TILT position for pipetting without holding the bottle
  • 100% bigger opening for easy pipetting
  • Tilted volume indicators for convenience
  • Vacuum resistant bottle for safe filter sterilisation
Biosafety cabinets and clean hoods are essential for working with cell cultures under sterile conditions. Unfortunately, they make theMore Product Information