Bottles, narrow or wide neck, square, with thread
Supplier: DWK Life Sciences
238102456 238162453 238163655 238164454 238165459 238202405 238203607 238204406 238262402 238263604 238264403 238265408 238103658
SCOT238102456EA 6.6 SGD
SCOT238102456 SCOT238162453 SCOT238163655 SCOT238164454 SCOT238165459 SCOT238202405 SCOT238203607 SCOT238204406 SCOT238262402 SCOT238263604 SCOT238264403 SCOT238265408 SCOT238103658
Bottles, narrow or wide neck, square, with thread
Bottles Glass Bottles
Soda-lime glass, clear or amber, without screw caps.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Closures in various designs are available on request
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials.