Weighing bottles, glass, DURAN®
Supplier: DWK Life Sciences
242111305 242112404 242101304 242102309 242103202 242104104 242112301 242111802
SCOT242111305EA 363 SGD
SCOT242111305 SCOT242112404 SCOT242101304 SCOT242102309 SCOT242103202 SCOT242104104 SCOT242112301 SCOT242111802
Weighing bottles, glass, DURAN®
Bottles Weighing Bottles
DURAN® Borosilicate glass 3.3, clear.
  • Interchangeable lids with ground joint
  • Writing area in white
  • Available in low or tall form
Order Now
SPECIFICATIONS
Environmentally Preferable