Jacketed coil condenser, DURAN®
Supplier: DWK Life Sciences
242557107 242557201 242558103 242558206
SCOT242557107EA 247 SGD
SCOT242557107 SCOT242557201 SCOT242558103 SCOT242558206
Jacketed coil condenser, DURAN®
Condensers
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cooling condensate from vapours, with cone and socket. Water supplied through plastic connectors.
This type of condenser has a very large heat exchange surface due to its coil condenser and the double jacket and is thus especially suited to working with low boiling point media.