Stirrer shafts, DURAN® KPG®
Supplier: DWK Life Sciences
245656409 245666701
SCOT245656409EA 75 SGD
SCOT245656409 SCOT245666701
Stirrer shafts, DURAN® KPG®
Stirrers Stirring Shafts
DURAN®, clear borosilicate glass 3.3.
  • Interchangeable
  • Polished surface
Order Now
SPECIFICATIONS
Environmentally Preferable