SBW glass stoppers, standard ground joint, octagonal
Supplier: DWK Life Sciences
246240604
SCOT246240604EA 111 SGD
SCOT246240604
SBW glass stoppers, standard ground joint, octagonal
Stoppers Glass Stoppers
These stoppers are made from SBW glass and have an octagonal head for good grip.
  • Solid form
  • Available amber or clear
Certifications: DIN 12252