Volumetric flasks, class A, amber, white markings, DURAN®
Supplier: DWK Life Sciences
246760954 246761059 246761256 246761453 246761753 246762458 246762552 246763257 246763651 246764459 246765455 246766357
SCOT246760954EA 54.9 SGD
SCOT246760954 SCOT246761059 SCOT246761256 SCOT246761453 SCOT246761753 SCOT246762458 SCOT246762552 SCOT246763257 SCOT246763651 SCOT246764459 SCOT246765455 SCOT246766357
Volumetric flasks, class A, amber, white markings, DURAN®
Flasks Volumetric Flasks
DURAN®, borosilicate glass 3.3, amber, with PE stopper, conformity certified.
  • White markings, with one graduation mark and new ergonomic polyethylene stopper
  • Certificate of conformity 'DE-M'
  • With batch certificate
*Wide neck.
With printed retrace code (batch identification).
Certifications: ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable