DURAN® Volumetric Flasks, Class A, Amber, White Markings
Supplier: DWK Life Sciences
246771257 246770955 246771454 246771754 246772459 246772553 246773258 246773652 246774451 246775456 246776358 246771051
SCOT246771257EA 180 SGD
SCOT246771257 SCOT246770955 SCOT246771454 SCOT246771754 SCOT246772459 SCOT246772553 SCOT246773258 SCOT246773652 SCOT246774451 SCOT246775456 SCOT246776358 SCOT246771051
DURAN® Volumetric Flasks, Class A, Amber, White Markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass 3.3, amber, with PE stopper, conformity certified.
  • White markings, with one graduation mark
  • Certificate of conformity 'DE-M' mark
  • Available with individual certificate
*Wide neck.
With printed retrace code (batch identification).