Desiccator bases, DURAN®
Supplier: DWK Life Sciences
247704604 247705703 247706108 247706605 247706905 247714605 247715704 247716109 247716606 247716906 247725705 247726101 247726607 247726907
SCOT247704604EA 129 SGD
SCOT247704604 SCOT247705703 SCOT247706108 SCOT247706605 SCOT247706905 SCOT247714605 SCOT247715704 SCOT247716109 SCOT247716606 SCOT247716906 SCOT247725705 SCOT247726101 SCOT247726607 SCOT247726907
Desiccator bases, DURAN®
Desiccators Desiccator Bases
DURAN®, clear borosilicate glass 3.3. Ideal for drying and drying processes, storage under vacuum and protection from the effects of air and moisture. Applications include cultivation of organisms, material research and corrosion testing, cleaning equipment for analysis of trace elements, gas and liquid reactions and they can be used as a mini cleanroom for chip technology.
  • Choice of various desiccator components to suit different applications
  • Increased safety due to DURAN® quality borosilicate glass
  • All models have a flat flange, except 467-0139 which
More Product Information