Stoppers, pointed, diamond-ground, DURAN®
Supplier: DWK Life Sciences
286117407 286118103 286117904 286113808 286114204 286114804 286115406 286116008 286116608
SCOT286117407EA 535 SGD
SCOT286117407 SCOT286118103 SCOT286117904 SCOT286113808 SCOT286114204 SCOT286114804 SCOT286115406 SCOT286116008 SCOT286116608
Stoppers, pointed, diamond-ground, DURAN®
Stoppers Glass Stoppers
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Pointed
  • Diamond-ground