Stoppers, flat, diamond-ground, DURAN®
Supplier: DWK Life Sciences
286121808 286122504 286123509 286124608 286126206 286129006 286130307 286130907 286127802
SCOT286121808EA 471 SGD
SCOT286121808 SCOT286122504 SCOT286123509 SCOT286124608 SCOT286126206 SCOT286129006 SCOT286130307 SCOT286130907 SCOT286127802
Stoppers, flat, diamond-ground, DURAN®
Stoppers Glass Stoppers
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Flat
  • Diamond-ground