Desiccator plates DURAN®
Supplier: DWK Life Sciences
290804606 290805705 290806101 290806607 290806907
SCOT290804606EA 16.4 SGD
SCOT290804606 SCOT290805705 SCOT290806101 SCOT290806607 SCOT290806907
Desiccator plates DURAN®
Desiccators Desiccators, Plates
Stainless steel 1.4301, Type 304, rust-free.