Rubber stopper, DURAN®
Supplier: DWK Life Sciences
292062803
SCOT292062803EA 84.9 SGD
SCOT292062803
Rubber stopper, DURAN®
Stoppers Rubber Stoppers
This bromobutyl rubber closure provides a gas-tight seal for all GL 25 and GL 45 laboratory bottles with sizes from 10 to 20000 ml.
  • Elastic material for reliable self-sealing after needle piercing
  • Upper surface features seven numbered hemispherical impressions for needle piercing
  • Resistant to temperature from –50 to +121 °C
Suitable for storing oxygen-sensitive materials, cultivation of anaerobic microorganisms (pressure resistent bottles recommended), aerobic methane oxidation (MOx) studies, storage of anaerobic culture media (amber glass bottlesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable