Screw cap, GL 45, PMP, DURAN® YOUTILITY®
Supplier: DWK Life Sciences
292292843
SCOT292292843EA 53.8 SGD
SCOT292292843
Screw cap, GL 45, PMP, DURAN® YOUTILITY®
Closures Bottle Closures
A highly transparent and ergonomic polymethylpentene cap for GL 45 DURAN® laboratory bottles.
  • High transparency (low haze) to visible light that is equivalent to DURAN® borosilicate glass
  • Compatible with all glass lab bottles and flasks with a GL 45 thread
  • Autoclavable at 121 or 134 °C, so is suitable for all GL 45 culture bottles and flasks
  • Retrace Code for traceability
  • Convenient liner-less YOUTILITY® cap design with ergonomic external grip
  • Safe with excellent chemical resistance - non toxic, BFA-free and halogen-free, food contact safe polymer (broadly
More Product Information