Over-caps, 45 mm, black phenolic, DURAN®
Supplier: DWK Life Sciences
292492901
SCOT292492901EA 40.3 SGD
SCOT292492901
Over-caps, 45 mm, black phenolic, DURAN®
Closures Bottle Closures
The 45 mm phenolic over-cap screws down over the open-top aluminium GL 45 cap covering and protecting the septum or stopper.
  • Made from high-quality phenol formaldehyde resin (PF)
  • High quality components
  • Recommended usable temperature is 150 °C
The phenolic cap protects the septum or stopper from damage and contamination during storage or shipping. Phenolic caps provide a wide range of chemical compatibility and temperature tolerance.