VWR®, Bottle carrying systems
Supplier: DWK Life Sciences
292807309
SCOT292807309EA 256 SGD
SCOT292807309
VWR®, Bottle carrying systems
Carriers Bottle Carriers/Containers
This modular bottle carrying system fits standard laboratory bottles that have a GL 45 neck finish.
  • Compatible with four bottle sizes 2, 5, 10 and 20 litres
  • All materials are fully autoclavable
  • Makes pouring liquids from the larger bottles more comfortable and safer
It is ergonomically designed and offers health and safety benefits to the user. It allows easy and safe transport of bottles around the laboratory or production facility.
Order Now
SPECIFICATIONS
Environmentally Preferable