GLS 80® wide mouth connection transfer cap system, DURAN®
Supplier: DWK Life Sciences
293109103
SCOT293109103EA 395 SGD
SCOT293109103
GLS 80® wide mouth connection transfer cap system, DURAN®
Closures Bottle Closures
PP and PTFE.
  • Flexible modular system
  • Very good temperature resistance
  • Safe transfer of liquid media within a closed and sterile system (evaporation is reduced)
Screw Cap GLS 80 with four ports (GL thread). Five different hose diameters (3,2; 6,0; 8,0; 10,00 and 12,0 mm) can be connected. Sterile pressure equalisation is possible through use of membrane filter. Unused ports can be provided with a blind cap.