Bottles, clear or amber glass, DURAN® PURE
Supplier: DWK Life Sciences
818062402 818063604 818061406 818011401 818012406 818013608 818014407 818015403 818016305 818017301 818018606 818019105 818064403 818065408 818066301 818067306 818068602 818069101
SCOT818062402EA 422 SGD
SCOT818062402 SCOT818063604 SCOT818061406 SCOT818011401 SCOT818012406 SCOT818013608 SCOT818014407 SCOT818015403 SCOT818016305 SCOT818017301 SCOT818018606 SCOT818019105 SCOT818064403 SCOT818065408 SCOT818066301 SCOT818067306 SCOT818068602 SCOT818069101
Bottles, clear or amber glass, DURAN® PURE
Bottles Glass Bottles
DURAN® PURE bottles clear or amber, with GL thread and dust cover. DURAN® PURE is a range of primary packaging products dedicated to the specific requirements and needs of the pharmaceutical industry.
  • Made from DURAN® borosilicate glass 3.3 Type I
  • Protective dust cover is applied in-line at the end of the manufacturing process while the glass still has a very high level of cleanness
  • Lot specific certificate of conformance is available via 'Retrace Code' internet download (includes USP, Ph. Eur., JP analysis per lot)
  • 100% lot traceability of DURAN® PURE
More Product Information