Accessories for continuous level control, SymLine®
Supplier: S.C.A.T.
106548
SCTV106548EA 0 SGD
SCTV106548
Accessories for continuous level control, SymLine®
Level Sensors
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
  • Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
  • Touchless alarm-resetting fields for two containers
  • Two switch inputs and two switch outputs for peripheral units
  • Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner,More Product Information