SafetyWasteCaps
Supplier: S.C.A.T.
107949 107960 107963 107966 107967 107969 108023 108024 108025 108026 108046 108047 108156 108200 108201 108205 108231 107034 107036 107037 107038 107029 107913 107915 107917 107922 107924 107926 107927 107928 107930 107935 107936 107942 107943 107947 107050 108055 107053 107052 107051 107257 107256 107259 107242 107246 108142 307912 307923 308921 307964 307500 308964 307925 307931 307944 308403 307916 307918 308401 308402 308404 308961 307108 307109 307961 107121 108143
SCTV107949EA 0 SGD
SCTV107949 SCTV107960 SCTV107963 SCTV107966 SCTV107967 SCTV107969 SCTV108023 SCTV108024 SCTV108025 SCTV108026 SCTV108046 SCTV108047 SCTV108156 SCTV108200 SCTV108201 SCTV108205 SCTV108231 SCTV107034 SCTV107036 SCTV107037 SCTV107038 SCTV107029 SCTV107913 SCTV107915 SCTV107917 SCTV107922 SCTV107924 SCTV107926 SCTV107927 SCTV107928 SCTV107930 SCTV107935 SCTV107936 SCTV107942 SCTV107943 SCTV107947 SCTV107050 SCTV108055 SCTV107053 SCTV107052 SCTV107051 SCTV107257 SCTV107256 SCTV107259 SCTV107242 SCTV107246 SCTV108142 SCTV307912 SCTV307923 SCTV308921 SCTV307964 SCTV307500 SCTV308964 SCTV307925 SCTV307931 SCTV307944 SCTV308403 SCTV307916 SCTV307918 SCTV308401 SCTV308402 SCTV308404 SCTV308961 SCTV307108 SCTV307109 SCTV307961 SCTV107121 SCTV108143
SafetyWasteCaps
Closures Safety Caps and Safety Waste Caps
The SCAT SafetyWasteCaps provide maximum safety for your HPLC liquid disposal. Safely dispose of your liquid laboratory waste without contaminating the ambient air by using SafetyWasteCaps with integrated exhaust air filters. These high-performance filters regulate the air pressure balance and prevent the escape of vapors due to the resulting overpressure in the disposal container. The variety of closures, connections and thread sizes allows optimal adaptation to your HPLC situation. Additional versions with grounding connections, level controls or safety funnels or offer every convenienceMore Product Information