Safety canisters
Supplier: S.C.A.T.
107950 107951 107952 107955 107956 107957 107958 107959 108020 108056 107707 107933 107710 107731 108115 108175 108189 108227
SCTV107950EA 58.7 SGD
SCTV107950 SCTV107951 SCTV107952 SCTV107955 SCTV107956 SCTV107957 SCTV107958 SCTV107959 SCTV108020 SCTV108056 SCTV107707 SCTV107933 SCTV107710 SCTV107731 SCTV108115 SCTV108175 SCTV108189 SCTV108227
Safety canisters
Safety Cans
These canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.
  • Made of very robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
  • Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
  • Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable