Tubing
Supplier: S.C.A.T.
108015 108016 108017 108018 108019 300021 300022 461053 461054 461055 461056
SCTV108015EA 0 SGD
SCTV108015 SCTV108016 SCTV108017 SCTV108018 SCTV108019 SCTV300021 SCTV300022 SCTV461053 SCTV461054 SCTV461055 SCTV461056
Tubing
Tubing Chromatography Tubing
Flexible tubing made of PP and PTFE-EL material.
  • Available in various materials, lengths and properties
Electrically conductive plastic tubing provides safe protection against ignition hazards and electrostatic charging.
Order Now
SPECIFICATIONS
Environmentally Preferable