Hand shovels for industry, Semadeni®
Supplier: SEMADENI
0039
SEMA0039EA 0 SGD
SEMA0039
Hand shovels for industry, Semadeni®
Scoops
These scoops are either made from PP or from LDPE.
  • Robust, break-proof and chemical resistant, even at lower temperatures
  • With solid handle