Bottles, wide neck, PE, rectangular, with screw cap
Supplier: SEMADENI
0063
SEMA0063EA 0 SGD
SEMA0063
Bottles, wide neck, PE, rectangular, with screw cap
Bottles Plastic Bottles
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Order Now
SPECIFICATIONS
Environmentally Preferable