Funnels, with short stem, PP
Supplier: SEMADENI
0221
SEMA0221EA 0 SGD
SEMA0221
Funnels, with short stem, PP
Funnels Filtration Funnels Filter Funnels
Funnels, made of PP, translucent, with slim stem.
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable