Magnetic stirring bars, with pivot ring, cylindrical
Supplier: SEMADENI
0278
SEMA0278EA 0 SGD
SEMA0278
Magnetic stirring bars, with pivot ring, cylindrical
Stirrers Magnetic Stirring Bars
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
  • Chemical resistant
  • Central ring helps in easier rotation
  • Temperature resistance up to 275 °C